CHIS5-08143

5-08146 5-08145 5-08143 5-08144
76234-508EA 131.02 USD
76234-508 76234-504 76234-506 76234-510
CHIS5-08143
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR