CHIS5-07991

5-07992 5-07991
76233-290EA 687.83 USD
76233-290 76233-288
CHIS5-07991
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR