CHIS5-07947

5-07950 5-07947 5-07949 5-07948
76234-382EA 262.03 USD
76234-382 76234-380 76233-250 76233-248
CHIS5-07947
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR