CHIS5-07829

5-07829 5-07830
76233-070EA 458.56 USD
76233-070 76233-068
CHIS5-07829
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR