CHIS5-07819

5-07819 5-07820
76233-050EA 360.29 USD
76233-050 76233-048
CHIS5-07819
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR