CHIS5-07775

5-07775 5-07776
76232-962EA 262.03 USD
76232-962 76232-960
CHIS5-07775
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR