CHIS5-07761

5-07761 5-07762
76234-302EA 232.55 USD
76234-302 76234-300
CHIS5-07761
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR