CHIS5-07747

5-07748 5-07747
76232-946EA 393.05 USD
76232-946 76232-944
CHIS5-07747
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR