CHIS5-07743

5-07743 5-07744
76232-938EA 196.52 USD
76232-938 76232-936
CHIS5-07743
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR