CHIS5-07739

5-07739 5-07740
76232-928EA 90.08 USD
76232-928 76232-930
CHIS5-07739
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR