CHIS5-07733

5-07734 5-07733
76232-918EA 409.43 USD
76232-918 76232-916
CHIS5-07733
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR