CHIS5-07719

5-07719 5-07720
76232-890EA 229.28 USD
76232-890 76232-888
CHIS5-07719
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR