CHIS5-07677

5-07677 5-07678
76232-838EA 294.79 USD
76232-838 76232-836
CHIS5-07677
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR