CHIS5-07673

5-07674 5-07673
76232-828EA 98.26 USD
76232-828 76232-830
CHIS5-07673
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR