CHIS5-07611

5-07612 5-07611
76232-714EA 196.52 USD
76232-714 76232-712
CHIS5-07611
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR