CHIS5-07601

5-07601 5-07602
76232-694EA 314.44 USD
76232-694 76232-692
CHIS5-07601
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR