CHIS5-07535

5-07536 5-07535
76232-594EA 466.74 USD
76232-594 76232-592
CHIS5-07535
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR