CHIS5-07517

5-07517 5-07518
76234-204EA 168.68 USD
76234-204 76234-206
CHIS5-07517
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR