CHIS5-07483

5-07484 5-07483
76232-522EA 343.92 USD
76232-522 76232-520
CHIS5-07483
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR