CHIS5-07469

5-07469 5-07470
76232-494EA 360.29 USD
76232-494 76232-492
CHIS5-07469
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR