CHIS5-07435

5-07436 5-07435
76232-450EA 458.56 USD
76232-450 76232-448
CHIS5-07435
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR