CHIS5-07393

5-07394 5-07393
76232-364EA 196.52 USD
76232-364 76232-366
CHIS5-07393
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR