CHIS5-07383

5-07383 5-07384
76232-346EA 573.2 USD
76232-346 76232-344
CHIS5-07383
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR