Mitomycin C

Warning

Synonyms: AmetycinMitomycin

2713-5 2713-25
76021-522EA 119.1 USD
76021-522 76021-524
Mitomycin C
Mitomycin C

Mitomycin C is an antitumor antibiotic isolated from Streptomyces sp. Mitomycin is activated by reduction in vivo generating oxygen radicals which produce crosslinks in DNA by alkylaion. This mechanism ultimately inhibits DNA synthesis which has a bacteriostatic effect. Mitomycin C has strong antitumor activity, especially against Ehrlich ascites tumor cells, and strong bactericidal action against gram-positive and gram-negative bacteria.

Formula: C₁₅H₁₈N₄O₅
MW: 334.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078109
CAS Number: 50-07-7
https://us.vwr.com/stibo/web/std.lang.all/22/31/23492231.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR