CHIS5-07325

5-07325 5-07326
76234-126EA 281.68 USD
76234-126 76234-124
CHIS5-07325
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR