CHIS5-07237

5-07237 5-07238
76234-092EA 245.66 USD
76234-092 76234-094
CHIS5-07237
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR