CHIS5-07229

5-07229 5-07230
76232-110EA 327.54 USD
76232-110 76232-108
CHIS5-07229
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR