CHIS5-07211

5-07212 5-07211
76232-074EA 352.11 USD
76232-074 76232-072
CHIS5-07211
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR