CHIS5-07187

5-07188 5-07187
76232-024EA 131.02 USD
76232-024 76232-026
CHIS5-07187
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR