CHIS5-07151

5-07152 5-07151
76231-962EA 327.54 USD
76231-962 76231-960
CHIS5-07151
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR