CHIS5-07093

5-07094 5-07093
76231-844EA 163.77 USD
76231-844 76231-846
CHIS5-07093
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR