CHIS5-07003

5-07004 5-07003
76231-688EA 171.95 USD
76231-688 76231-690
CHIS5-07003
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR