CHIS5-06943

5-06944 5-06943
76231-584EA 168.68 USD
76231-584 76231-586
CHIS5-06943
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR