CHIS5-06929

5-06930 5-06929
76231-556EA 58.96 USD
76231-556 76231-558
CHIS5-06929
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR