CHIS5-06811

5-06811 5-06812
76233-880EA 40.95 USD
76233-880 76233-882
CHIS5-06811
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR