CHIS5-06785

5-06786 5-06785
76231-444EA 131.02 USD
76231-444 76231-446
CHIS5-06785
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR