CHIS5-06767

5-06768 5-06767
76231-408EA 65.51 USD
76231-408 76231-410
CHIS5-06767
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR