CHIS5-06753

5-06754 5-06753
76231-380EA 245.66 USD
76231-380 76231-382
CHIS5-06753
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR