CHIS5-06749

5-06750 5-06749
76231-372EA 65.51 USD
76231-372 76231-374
CHIS5-06749
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR