CHIS5-06701

5-06701 5-06702
76233-822EA 663.27 USD
76233-822 76233-820
CHIS5-06701
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR