CHIS5-06523

5-06524 5-06523
76231-060EA 548.63 USD
76231-060 76231-062
CHIS5-06523
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR