CHIS5-06501

5-06501 5-06502
76231-016EA 65.51 USD
76231-016 76231-018
CHIS5-06501
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR