CHIS5-06441

5-06441 5-06442
76230-906EA 818.85 USD
76230-906 76230-904
CHIS5-06441
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR