CHIS5-06435

5-06435 5-06436
76230-894EA 610.86 USD
76230-894 76230-892
CHIS5-06435
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR