CHIS5-06299

5-06299 5-06300
76230-678EA 818.85 USD
76230-678 76230-676
CHIS5-06299
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR