CHIS5-06203

5-06204 5-06203
76230-518EA 327.54 USD
76230-518 76230-516
CHIS5-06203
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR