CHIS5-06177

5-06178 5-06177
76230-480EA 294.79 USD
76230-480 76230-482
CHIS5-06177
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR