CHIS5-06165

5-06166 5-06165
76230-458EA 311.16 USD
76230-458 76230-456
CHIS5-06165
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR