CHIS5-06161

5-06162 5-06161
76230-448EA 229.28 USD
76230-448 76230-450
CHIS5-06161
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR