CHIS5-06119

5-06120 5-06119
76230-374EA 119.55 USD
76230-374 76230-372
CHIS5-06119
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR